Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTPBP4
Entrez Gene ID
23560
Gene Name
GTP binding protein 4
Gene Aliases
CRFG, NGB, NOG1
Chromosome Location
Chr.10: 988409 - 1017768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012341.2 NP_036473.2 8 - 9 979 86
GenBank mRNA AF120334.1 - 8 - 9 931 86
AF325353.1 - 8 - 9 908 86
AK001548.1 - 8 - 9 926 86
AK001552.1 - 8 - 9 981 86
AK097093.1 - 6 - 7 1787 86
AK295231.1 - 7 - 8 822 86
AK301721.1 - 8 - 9 1094 86
AK302219.1 - 7 - 8 784 86
BC033784.1 - 8 - 9 902 86
BC038975.2 - 8 - 9 940 86

View Full Product Details