The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012456.1 | NP_001012474.1 | 2 - 3 | 178 | 156 |
NM_014302.3 | NP_055117.1 | 2 - 3 | 182 | 156 | |
GenBank mRNA | AF054184.1 | - | 2 - 3 | 177 | 156 |
AF086539.1 | - | 2 - 3 | 138 | 156 | |
BC009480.2 | - | 2 - 3 | 120 | 156 | |
BC051840.1 | - | - | 102 | 156 | |
BU932672.1 | - | - | 181 | 156 | |
CD385140.1 | - | 2 - 3 | 156 | 156 |