The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182497.3 | NP_872303.2 | 6 - 7 | 1007 | 68 |
XM_017024189.1 | XP_016879678.1 | 4 - 5 | 2774 | 68 | |
GenBank mRNA | AJ786658.1 | - | 4 - 5 | 847 | 68 |
AK093919.1 | - | 6 - 7 | 68 | ||
BC130396.1 | - | 5 - 6 | 882 | 68 | |
BC144250.1 | - | 5 - 6 | 882 | 68 | |
BC150174.1 | - | 6 - 7 | 1007 | 68 | |
BC150191.1 | - | 6 - 7 | 1007 | 68 |