The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004304.4 | NP_004295.2 | 22 - 23 | 4468 | 75 |
GenBank mRNA | AB209477.4 | - | 22 - 23 | 4005 | 75 |
EU788003.1 | - | 22 - 23 | 4423 | 75 | |
EU788004.1 | - | 22 - 23 | 4423 | 75 | |
EU788009.1 | - | 22 - 23 | 4423 | 75 | |
EU788010.1 | - | 22 - 23 | 4423 | 75 | |
FJ838786.1 | - | 22 - 23 | 4423 | 75 | |
FJ838787.1 | - | 22 - 23 | 4423 | 75 | |
GU128152.1 | - | 22 - 23 | 3516 | 75 | |
GU128154.1 | - | 22 - 23 | 3516 | 75 | |
GU128155.1 | - | 22 - 23 | 3516 | 75 | |
GU128156.1 | - | 22 - 23 | 3516 | 75 | |
GU128157.1 | - | 22 - 23 | 3516 | 75 | |
LN864494.1 | - | 3 - 4 | 716 | 75 | |
U62540.1 | - | 22 - 23 | 4427 | 75 | |
U66559.1 | - | 22 - 23 | 3664 | 75 |