The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018060.3 | NP_060530.3 | 16 - 17 | 2150 | 66 |
GenBank mRNA | AK001188.1 | - | 14 - 15 | 1625 | 66 |
AK001726.1 | - | 10 - 11 | 1138 | 66 | |
AK022665.1 | - | 6 - 7 | 717 | 66 | |
AK291432.1 | - | 16 - 17 | 1959 | 66 | |
AY267462.1 | - | 16 - 17 | 2161 | 66 | |
BC010218.1 | - | 12 - 13 | 1330 | 66 | |
BC040376.1 | - | 16 - 17 | 1883 | 66 | |
BC047880.1 | - | 15 - 16 | 1677 | 66 | |
BC137438.1 | - | 16 - 17 | 2120 | 66 | |
D28500.1 | - | 16 - 17 | 1991 | 66 |