The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018060.3 | NP_060530.3 | 19 - 20 | 2516 | 110 |
GenBank mRNA | AK001188.1 | - | 17 - 18 | 1991 | 110 |
AK001726.1 | - | 13 - 14 | 1504 | 110 | |
AK022665.1 | - | 9 - 10 | 1083 | 110 | |
AK291432.1 | - | 19 - 20 | 2325 | 110 | |
AY267462.1 | - | 19 - 20 | 2527 | 110 | |
BC010218.1 | - | 15 - 16 | 1696 | 110 | |
BC040376.1 | - | 19 - 20 | 2249 | 110 | |
BC047880.1 | - | 18 - 19 | 2043 | 110 | |
BC137438.1 | - | 19 - 20 | 2486 | 110 | |
D28500.1 | - | 19 - 20 | 2357 | 110 |