The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018060.3 | NP_060530.3 | 21 - 22 | 2858 | 75 |
GenBank mRNA | AK001188.1 | - | 19 - 20 | 2332 | 75 |
AK001726.1 | - | 15 - 16 | 1846 | 75 | |
AK291432.1 | - | 21 - 22 | 2667 | 75 | |
AY267462.1 | - | 21 - 22 | 2869 | 75 | |
BC010218.1 | - | 17 - 18 | 2038 | 75 | |
BC040376.1 | - | 21 - 22 | 2591 | 75 | |
BC047880.1 | - | 20 - 21 | 2385 | 75 | |
BC137438.1 | - | 21 - 22 | 2828 | 75 | |
D28500.1 | - | 21 - 22 | 2699 | 75 |