The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018060.3 | NP_060530.3 | 6 - 7 | 967 | 142 |
GenBank mRNA | AK001188.1 | - | 4 - 5 | 442 | 142 |
AK291432.1 | - | 6 - 7 | 776 | 142 | |
AY267462.1 | - | 6 - 7 | 978 | 142 | |
BC010218.1 | - | 2 - 3 | 147 | 142 | |
BC040376.1 | - | 6 - 7 | 700 | 142 | |
BC047880.1 | - | 5 - 6 | 494 | 142 | |
BC137438.1 | - | 6 - 7 | 937 | 142 | |
D28500.1 | - | 6 - 7 | 808 | 142 |