Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMG1
Entrez Gene ID
10436
Gene Name
EMG1, N1-specific pseudouridine methyltransferase
Gene Aliases
C2F, Grcc2f, NEP1
Chromosome Location
Chr.12: 6970781 - 6997428 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320049.1 NP_001306978.1 2 - 3 412 152
NM_006331.7 NP_006322.4 2 - 3 412 152
NR_135131.1 2 - 3 412 152
GenBank mRNA AA314580.1 - 2 - 3 260 152
AK291597.1 - 2 - 3 277 152
AK298207.1 - 2 - 3 280 152
AK298745.1 - 2 - 3 272 152
BC055314.1 - 2 - 3 304 152
CD704778.1 - 2 - 3 401 152
DA070200.1 - 2 - 3 362 152
DC403138.1 - 2 - 3 412 152
U72514.1 - 2 - 3 255 152

View Full Product Details