Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDR1
Entrez Gene ID
780
Gene Name
discoidin domain receptor tyrosine kinase 1
Gene Aliases
CAK, CD167, DDR, EDDR1, HGK2, MCK10, NEP, NTRK4, PTK3, PTK3A, RTK6, TRKE
Chromosome Location
Chr.6: 30880909 - 30900156 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202521.1 NP_001189450.1 7 - 8 1146 83
NM_001202522.1 NP_001189451.1 7 - 8 1146 83
NM_001202523.1 NP_001189452.1 8 - 9 1398 83
NM_001297652.1 NP_001284581.1 8 - 9 1517 83
NM_001297653.1 NP_001284582.1 8 - 9 1319 83
NM_001297654.1 NP_001284583.1 8 - 9 1319 83
NM_001954.4 NP_001945.3 10 - 11 1437 83
NM_013993.2 NP_054699.2 9 - 10 1363 83
NM_013994.2 NP_054700.2 7 - 8 1146 83
XM_006715185.2 XP_006715248.1 8 - 9 1310 83
XM_011514882.2 XP_011513184.1 8 - 9 1310 83
XM_011514883.1 XP_011513185.1 10 - 11 1425 83
XM_011514884.1 XP_011513186.1 10 - 11 1501 83
XM_011514885.1 XP_011513187.1 8 - 9 1348 83
XM_011514886.1 XP_011513188.1 8 - 9 1213 83
XM_011514887.1 XP_011513189.1 8 - 9 1363 83
XM_011514888.2 XP_011513190.1 8 - 9 1310 83
XM_017011268.1 XP_016866757.1 8 - 9 1180 83
XM_017011269.1 XP_016866758.1 8 - 9 1363 83
GenBank mRNA AB210021.1 - 9 - 10 1258 83
AK130776.1 - 3 - 4 492 83
AK291621.1 - 8 - 9 1212 83
AK294793.1 - 8 - 9 1384 83
BC008716.2 - 10 - 11 1437 83
BC013400.2 - 10 - 11 1437 83
BC070070.1 - 8 - 9 1208 83
L11315.1 - 9 - 10 1245 83
L20817.1 - 8 - 9 1296 83
L57508.1 - 8 - 9 1296 83
X74979.1 - 7 - 8 1149 83
Z29093.1 - 7 - 8 1440 83

View Full Product Details