Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDR1
Entrez Gene ID
780
Gene Name
discoidin domain receptor tyrosine kinase 1
Gene Aliases
CAK, CD167, DDR, EDDR1, HGK2, MCK10, NEP, NTRK4, PTK3, PTK3A, RTK6, TRKE
Chromosome Location
Chr.6: 30880909 - 30900156 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202521.1 NP_001189450.1 1 - 2 124 80
NM_001202522.1 NP_001189451.1 1 - 2 124 80
NM_001202523.1 NP_001189452.1 2 - 3 376 80
NM_001297652.1 NP_001284581.1 2 - 3 495 80
NM_001297653.1 NP_001284582.1 2 - 3 297 80
NM_001297654.1 NP_001284583.1 2 - 3 297 80
NM_001954.4 NP_001945.3 4 - 5 415 80
NM_013993.2 NP_054699.2 3 - 4 341 80
NM_013994.2 NP_054700.2 1 - 2 124 80
XM_006715185.2 XP_006715248.1 2 - 3 288 80
XM_011514882.2 XP_011513184.1 2 - 3 288 80
XM_011514883.1 XP_011513185.1 4 - 5 403 80
XM_011514884.1 XP_011513186.1 4 - 5 479 80
XM_011514885.1 XP_011513187.1 2 - 3 326 80
XM_011514886.1 XP_011513188.1 2 - 3 191 80
XM_011514887.1 XP_011513189.1 2 - 3 341 80
XM_011514888.2 XP_011513190.1 2 - 3 288 80
XM_017011268.1 XP_016866757.1 2 - 3 158 80
XM_017011269.1 XP_016866758.1 2 - 3 341 80
GenBank mRNA AB210021.1 - 3 - 4 236 80
AK291621.1 - 2 - 3 190 80
AK294793.1 - 2 - 3 362 80
AK295643.1 - 2 - 3 189 80
BC008716.2 - 4 - 5 415 80
BC013400.2 - 4 - 5 415 80
BC070070.1 - 2 - 3 186 80
CN362335.1 - 3 - 4 341 80
DA461082.1 - 2 - 3 496 80
DC369107.1 - 2 - 3 376 80
EU826613.1 - 1 - 2 82 80
EU826614.1 - 1 - 2 82 80
L11315.1 - 3 - 4 80
L20817.1 - 2 - 3 274 80
L57508.1 - 2 - 3 274 80
X74979.1 - 1 - 2 127 80
Z29093.1 - 1 - 2 418 80

View Full Product Details