The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020546.2 | NP_065433.2 | 8 - 9 | 1337 | 65 |
XM_011513942.2 | XP_011512244.1 | 8 - 9 | 1284 | 65 | |
GenBank mRNA | AB028983.1 | - | 5 - 6 | 658 | 65 |
AF410885.1 | - | 8 - 9 | 1296 | 65 | |
AK123013.1 | - | 7 - 8 | 1238 | 65 | |
AK127036.1 | - | 8 - 9 | 1285 | 65 | |
AK294555.1 | - | 5 - 6 | 793 | 65 | |
AK295531.1 | - | 5 - 6 | 677 | 65 | |
AK299775.1 | - | 7 - 8 | 817 | 65 | |
AK304041.1 | - | 8 - 9 | 1172 | 65 | |
BC111743.1 | - | 8 - 9 | 1275 | 65 |