The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021924.4 | NP_068743.2 | 12 - 13 | 1544 | 95 |
GenBank mRNA | AF258676.1 | - | 13 - 14 | 1700 | 95 |
AF301909.1 | - | 12 - 13 | 1379 | 95 | |
AK000226.1 | - | 12 - 13 | 1562 | 95 | |
AK025012.1 | - | 5 - 6 | 532 | 96 | |
AK308040.1 | - | 12 - 13 | 1468 | 95 |