The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171968.1 | NP_001165439.1 | 1 - 2 | 246 | 115 |
NM_021924.4 | NP_068743.2 | 1 - 2 | 246 | 115 | |
NM_031264.3 | NP_112554.2 | 1 - 2 | 246 | 115 | |
XM_006718253.3 | XP_006718316.1 | 1 - 2 | 146 | 115 | |
XM_011520188.2 | XP_011518490.1 | 1 - 2 | 146 | 115 | |
XM_011520189.2 | XP_011518491.1 | 1 - 2 | 146 | 115 | |
XM_011520190.2 | XP_011518492.1 | 1 - 2 | 146 | 115 | |
XM_011520191.2 | XP_011518493.1 | 1 - 2 | 146 | 115 | |
GenBank mRNA | AF258675.1 | - | 2 - 3 | 402 | 115 |
AF258676.1 | - | 2 - 3 | 402 | 115 | |
AF258677.1 | - | 2 - 3 | 303 | 115 | |
AF258678.1 | - | 1 - 2 | 345 | 115 | |
AF276242.1 | - | 1 - 2 | 81 | 115 | |
AF301909.1 | - | 1 - 2 | 81 | 115 | |
AK000226.1 | - | 1 - 2 | 111 | 115 | |
AK000384.1 | - | 1 - 2 | 151 | 115 | |
AK300991.1 | - | 2 - 3 | 94 | ||
AK308040.1 | - | 1 - 2 | 121 | 115 | |
AK309157.1 | - | 1 - 2 | 134 | 115 | |
AY358368.1 | - | 1 - 2 | 103 | 115 | |
BC038405.1 | - | 1 - 2 | 103 | 115 | |
DA373160.1 | - | 1 - 2 | 188 | 115 | |
DQ491193.1 | - | 1 - 2 | 153 | 115 |