The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080439.2 | NP_001073908.2 | 1 - 2 | 680 | 126 |
XM_011524283.1 | XP_011522585.1 | 1 - 2 | 672 | 126 | |
GenBank mRNA | BC033017.1 | - | 1 - 2 | 644 | 126 |
BC033020.1 | - | 1 - 2 | 667 | 126 |