The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182628.2 | NP_872434.2 | 5 - 6 | 513 | 77 |
XM_006713623.2 | XP_006713686.1 | 6 - 7 | 664 | 77 | |
XM_017006320.1 | XP_016861809.1 | 6 - 7 | 664 | 77 | |
XM_017006321.1 | XP_016861810.1 | 6 - 7 | 664 | 77 | |
XM_017006322.1 | XP_016861811.1 | 6 - 7 | 664 | 77 | |
XM_017006323.1 | XP_016861812.1 | 6 - 7 | 664 | 77 | |
XM_017006324.1 | XP_016861813.1 | 6 - 7 | 664 | 77 | |
XM_017006325.1 | XP_016861814.1 | 6 - 7 | 664 | 77 | |
XM_017006326.1 | XP_016861815.1 | 6 - 7 | 664 | 77 | |
XM_017006327.1 | XP_016861816.1 | 6 - 7 | 664 | 77 | |
GenBank mRNA | AK097402.1 | - | 5 - 6 | 513 | 77 |
BC101367.2 | - | 4 - 5 | 443 | 77 | |
BC101368.1 | - | 4 - 5 | 443 | 77 | |
BC101369.1 | - | 4 - 5 | 443 | 77 |