Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SENP1
Entrez Gene ID
29843
Gene Name
SUMO1/sentrin specific peptidase 1
Gene Aliases
SuPr-2
Chromosome Location
Chr.12: 48042893 - 48106308 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267594.1 NP_001254523.1 17 - 18 2202 115
NM_001267595.1 NP_001254524.1 17 - 18 2457 115
NR_051991.1 18 - 19 2643 115
NR_051992.1 18 - 19 2656 115
XM_006719361.3 XP_006719424.1 17 - 18 2337 115
XM_006719362.2 XP_006719425.1 18 - 19 2109 115
XM_011538244.2 XP_011536546.1 16 - 17 2338 115
XM_011538245.2 XP_011536547.1 16 - 17 2289 115
XM_017019229.1 XP_016874718.1 18 - 19 2475 115
XM_017019230.1 XP_016874719.1 17 - 18 2391 115
XM_017019231.1 XP_016874720.1 18 - 19 2375 115
XM_017019232.1 XP_016874721.1 18 - 19 2102 115
XM_017019233.1 XP_016874722.1 17 - 18 2385 115
XM_017019234.1 XP_016874723.1 17 - 18 2295 115
XM_017019235.1 XP_016874724.1 17 - 18 2028 115
XM_017019236.1 XP_016874725.1 16 - 17 2263 115
XM_017019237.1 XP_016874726.1 16 - 17 2163 115
XM_017019238.1 XP_016874727.1 18 - 19 2413 115
XM_017019239.1 XP_016874728.1 17 - 18 2453 115
XM_017019240.1 XP_016874729.1 17 - 18 2374 115
GenBank mRNA AF149770.1 - 15 - 16 1874 115
AK292060.1 - 17 - 18 1973 115
AK312874.1 - 17 - 18 2203 115
BC045639.1 - 17 - 18 2273 115
BX537920.1 - 18 - 19 2656 115
BX640784.1 - 18 - 19 2487 115

View Full Product Details