The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033553.2 | NP_001028725.1 | 11 - 12 | 2886 | 86 |
NM_001033555.2 | NP_001028727.1 | 9 - 10 | 2795 | 86 | |
NM_001243439.1 | NP_001230368.1 | 11 - 12 | 2945 | 86 | |
XM_005256860.2 | XP_005256917.1 | 8 - 9 | 1215 | 86 | |
XM_017025317.1 | XP_016880806.1 | 10 - 11 | 2862 | 86 | |
XM_017025318.1 | XP_016880807.1 | 10 - 11 | 2862 | 86 | |
XM_017025319.1 | XP_016880808.1 | 11 - 12 | 3004 | 86 | |
XM_017025320.1 | XP_016880809.1 | 11 - 12 | 2980 | 86 | |
XM_017025321.1 | XP_016880810.1 | 9 - 10 | 2819 | 86 | |
GenBank mRNA | AK295093.1 | - | 11 - 12 | 2918 | 86 |
AK295811.1 | - | 8 - 9 | 2422 | 86 | |
AK298601.1 | - | 8 - 9 | 1627 | 86 | |
AK304186.1 | - | 9 - 10 | 2717 | 86 | |
AY816327.1 | - | 9 - 10 | 2700 | 86 | |
AY816329.1 | - | 11 - 12 | 2886 | 86 | |
BC021123.1 | - | 9 - 10 | 2693 | 86 | |
CN267630.1 | - | 3 - 4 | 261 | 86 |