Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPECC1
Entrez Gene ID
92521
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1
Gene Aliases
CYTSB, HCMOGT-1, HCMOGT1, NSP
Chromosome Location
Chr.17: 20009301 - 20319026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033553.2 NP_001028725.1 12 - 13 2988 64
NM_001033555.2 NP_001028727.1 10 - 11 2897 64
NM_001243439.1 NP_001230368.1 12 - 13 3047 64
XM_005256860.2 XP_005256917.1 9 - 10 1317 64
XM_017025317.1 XP_016880806.1 11 - 12 2964 64
XM_017025319.1 XP_016880808.1 12 - 13 3106 64
XM_017025321.1 XP_016880810.1 10 - 11 2921 64
GenBank mRNA AK295093.1 - 12 - 13 3020 64
AK295811.1 - 9 - 10 2524 64
AK298601.1 - 9 - 10 1729 64
AK301318.1 - 9 - 10 1239 64
AK304186.1 - 10 - 11 2819 64
AY816327.1 - 10 - 11 2802 64
AY816329.1 - 12 - 13 2988 64
BC021123.1 - 10 - 11 2795 64
CN267630.1 - 4 - 5 363 64

View Full Product Details