Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CACNA2D4
Entrez Gene ID
93589
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 4
Gene Aliases
RCD4
Chromosome Location
Chr.12: 1791957 - 1922606 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172364.4 NP_758952.4 17 - 18 2039 78
GenBank mRNA AF516695.1 - 17 - 18 2171 78
BC150186.1 - 17 - 18 2024 78
BX537436.1 - 17 - 18 2002 78
BX537437.1 - 17 - 18 2040 78

View Full Product Details