The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172364.4 | NP_758952.4 | 1 - 2 | 462 | 58 |
XM_011521041.2 | XP_011519343.1 | 1 - 2 | 4364 | 58 | |
GenBank mRNA | AF516695.1 | - | 1 - 2 | 594 | 58 |
BC150186.1 | - | 1 - 2 | 395 | 58 | |
BX537436.1 | - | 1 - 2 | 425 | 58 | |
BX537437.1 | - | 1 - 2 | 463 | 58 |