Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CACNA2D4
Entrez Gene ID
93589
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 4
Gene Aliases
RCD4
Chromosome Location
Chr.12: 1791957 - 1922606 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172364.4 NP_758952.4 32 - 33 3151 90
XM_011521041.2 XP_011519343.1 31 - 32 6990 90
GenBank mRNA AF516695.1 - 32 - 33 3283 90
AK301234.1 - 5 - 6 392 90
AL137658.1 - 9 - 10 690 90
BC048288.1 - 4 - 5 257 90
BC150186.1 - 31 - 32 3074 90
BI908421.1 - 5 - 6 358 90
BX537437.1 - 32 - 33 3152 90

View Full Product Details