Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CACNA2D4
Entrez Gene ID
93589
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 4
Gene Aliases
RCD4
Chromosome Location
Chr.12: 1791957 - 1922606 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172364.4 NP_758952.4 33 - 34 3206 59
XM_011521041.2 XP_011519343.1 32 - 33 7045 59
GenBank mRNA AF516695.1 - 33 - 34 3338 59
AK301234.1 - 6 - 7 447 59
AL137658.1 - 10 - 11 745 59
BC048288.1 - 5 - 6 312 59
BC150186.1 - 32 - 33 3129 59
BI908421.1 - 6 - 7 60
BX537437.1 - 33 - 34 3207 59

View Full Product Details