The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172364.4 | NP_758952.4 | 3 - 4 | 651 | 56 |
XM_011521041.2 | XP_011519343.1 | 3 - 4 | 4553 | 56 | |
GenBank mRNA | AF516695.1 | - | 3 - 4 | 783 | 56 |
BC150186.1 | - | 3 - 4 | 584 | 56 | |
BX537436.1 | - | 3 - 4 | 614 | 56 | |
BX537437.1 | - | 3 - 4 | 652 | 56 |