The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207014.1 | NP_001193943.1 | 2 - 3 | 308 | 66 |
XM_011545327.1 | XP_011543629.1 | 2 - 3 | 77 | 66 | |
GenBank mRNA | AK122936.1 | - | 1 - 2 | 1401 | 66 |
AK290124.1 | - | 2 - 3 | 245 | 66 | |
AK309905.1 | - | 2 - 3 | 111 | 66 | |
DC398178.1 | - | 2 - 3 | 309 | 66 |