Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FGF11
Entrez Gene ID
2256
Gene Name
fibroblast growth factor 11
Gene Aliases
FGF-11, FHF-3, FHF3
Chromosome Location
Chr.17: 7438273 - 7444932 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303460.1 NP_001290389.1 4 - 5 508 151
NM_004112.3 NP_004103.1 4 - 5 1200 151
NR_130156.1 4 - 5 671 151
GenBank mRNA AK293131.1 - 4 - 5 508 151
AK308812.1 - 4 - 5 671 151
BC032502.1 - 4 - 5 857 151
BC108265.1 - 4 - 5 1200 151

View Full Product Details