The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300743.1 | NP_001287672.1 | 19 - 20 | 1901 | 70 |
NM_020314.5 | NP_064710.4 | 21 - 22 | 2102 | 70 | |
XM_005255435.3 | XP_005255492.1 | 21 - 22 | 2054 | 70 | |
XM_006721065.3 | XP_006721128.1 | 19 - 20 | 1853 | 70 | |
XM_011545896.2 | XP_011544198.1 | 21 - 22 | 1803 | 70 | |
XM_011545897.2 | XP_011544199.1 | 14 - 15 | 1170 | 70 | |
GenBank mRNA | AF461052.2 | - | 15 - 16 | 1598 | 70 |
AK024693.1 | - | 17 - 18 | 1372 | 70 | |
AK095078.1 | - | 19 - 20 | 1598 | 70 | |
AK290286.1 | - | 21 - 22 | 1799 | 70 | |
AL136744.1 | - | 8 - 9 | 715 | 70 | |
AL832159.1 | - | 21 - 22 | 1798 | 70 | |
BC014900.2 | - | 20 - 21 | 1770 | 70 | |
BC050464.1 | - | 21 - 22 | 1842 | 70 | |
BC058845.1 | - | 21 - 22 | 1801 | 70 | |
BX537867.1 | - | 21 - 22 | 1814 | 70 |