Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF2
Entrez Gene ID
9181
Gene Name
Rho/Rac guanine nucleotide exchange factor 2
Gene Aliases
GEF, GEF-H1, GEFH1, LFP40, P40
Chromosome Location
Chr.1: 155946839 - 155991263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162383.1 NP_001155855.1 13 - 14 1832 82
NM_001162384.1 NP_001155856.1 13 - 14 1829 82
NM_004723.3 NP_004714.2 13 - 14 1757 82
XM_005245587.2 XP_005245644.1 13 - 14 1885 82
XM_005245588.2 XP_005245645.1 13 - 14 1882 82
XM_005245589.4 XP_005245646.1 13 - 14 2038 82
XM_005245590.3 XP_005245647.1 13 - 14 1760 82
XM_005245591.3 XP_005245648.1 14 - 15 1876 82
XM_005245592.2 XP_005245649.1 13 - 14 2029 82
XM_005245593.4 XP_005245650.1 13 - 14 1760 82
XM_005245594.2 XP_005245651.1 13 - 14 2746 82
XM_006711622.3 XP_006711685.1 13 - 14 3023 82
XM_006711623.3 XP_006711686.1 12 - 13 1759 82
XM_006711624.3 XP_006711687.1 12 - 13 1786 82
XM_006711625.3 XP_006711688.1 12 - 13 1692 82
XM_006711626.2 XP_006711689.1 12 - 13 2678 82
XM_011510136.2 XP_011508438.2 17 - 18 2441 82
XM_011510137.2 XP_011508439.2 17 - 18 2444 82
XM_011510139.2 XP_011508441.1 17 - 18 2444 82
XM_011510141.2 XP_011508443.1 13 - 14 2562 82
XM_011510143.2 XP_011508445.1 9 - 10 1308 82
XM_017002794.1 XP_016858283.1 17 - 18 2441 82
XM_017002795.1 XP_016858284.1 13 - 14 1776 82
XM_017002796.1 XP_016858285.1 13 - 14 3020 82
XM_017002797.1 XP_016858286.1 13 - 14 1760 82
XM_017002798.1 XP_016858287.1 12 - 13 2955 82
XM_017002799.1 XP_016858288.1 12 - 13 1756 82
GenBank mRNA AB014551.3 - 17 - 18 2100 82
AF486838.1 - 13 - 14 1711 82
AL512715.1 - 13 - 14 1742 82
BC020567.1 - 13 - 14 1814 82
BT007407.1 - 12 - 13 1633 82
U72206.1 - 13 - 14 1819 82

View Full Product Details