Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ARHGEF2
- Entrez Gene ID
-
9181
- Gene Name
- Rho/Rac guanine nucleotide exchange factor 2
- Gene Aliases
-
GEF, GEF-H1, GEFH1, LFP40, P40
- Chromosome Location
-
Chr.1: 155946839 - 155991263 on Build GRCh38
View Full Product Details