Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGEF2
Entrez Gene ID
9181
Gene Name
Rho/Rac guanine nucleotide exchange factor 2
Gene Aliases
GEF, GEF-H1, GEFH1, LFP40, P40
Chromosome Location
Chr.1: 155946839 - 155991263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162383.1 NP_001155855.1 18 - 19 2325 83
NM_001162384.1 NP_001155856.1 18 - 19 2322 83
NM_004723.3 NP_004714.2 18 - 19 2250 83
XM_005245587.2 XP_005245644.1 18 - 19 2378 83
XM_005245588.2 XP_005245645.1 18 - 19 2375 83
XM_005245589.4 XP_005245646.1 18 - 19 2531 83
XM_005245590.3 XP_005245647.1 18 - 19 2253 83
XM_005245591.3 XP_005245648.1 19 - 20 2369 83
XM_005245592.2 XP_005245649.1 18 - 19 2522 83
XM_005245593.4 XP_005245650.1 18 - 19 2253 83
XM_005245594.2 XP_005245651.1 18 - 19 3239 83
XM_006711622.3 XP_006711685.1 18 - 19 3516 83
XM_006711623.3 XP_006711686.1 17 - 18 2252 83
XM_006711624.3 XP_006711687.1 17 - 18 2279 83
XM_006711625.3 XP_006711688.1 17 - 18 2185 83
XM_006711626.2 XP_006711689.1 17 - 18 3171 83
XM_011510136.2 XP_011508438.2 22 - 23 2934 83
XM_011510137.2 XP_011508439.2 22 - 23 2937 83
XM_011510139.2 XP_011508441.1 22 - 23 2937 83
XM_011510141.2 XP_011508443.1 18 - 19 3055 83
XM_011510143.2 XP_011508445.1 14 - 15 1801 83
XM_017002794.1 XP_016858283.1 22 - 23 2934 83
XM_017002795.1 XP_016858284.1 18 - 19 2269 83
XM_017002796.1 XP_016858285.1 18 - 19 3513 83
XM_017002797.1 XP_016858286.1 18 - 19 2253 83
XM_017002798.1 XP_016858287.1 17 - 18 3448 83
XM_017002799.1 XP_016858288.1 17 - 18 2249 83
GenBank mRNA AB014551.3 - 22 - 23 2593 83
AF486838.1 - 18 - 19 2204 83
AL512715.1 - 18 - 19 2235 83
BC020567.1 - 18 - 19 2307 83
BT007407.1 - 17 - 18 2126 83
U72206.1 - 18 - 19 2309 83
X15610.1 - 4 - 5 317 83

View Full Product Details