Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF2
Entrez Gene ID
9181
Gene Name
Rho/Rac guanine nucleotide exchange factor 2
Gene Aliases
GEF, GEF-H1, GEFH1, LFP40, P40
Chromosome Location
Chr.1: 155946839 - 155991263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162383.1 NP_001155855.1 7 - 8 842 64
NM_001162384.1 NP_001155856.1 7 - 8 839 64
NM_004723.3 NP_004714.2 7 - 8 767 64
XM_005245587.2 XP_005245644.1 7 - 8 895 64
XM_005245588.2 XP_005245645.1 7 - 8 892 64
XM_005245589.4 XP_005245646.1 7 - 8 1048 64
XM_005245590.3 XP_005245647.1 7 - 8 770 64
XM_005245591.3 XP_005245648.1 8 - 9 886 64
XM_005245592.2 XP_005245649.1 7 - 8 1039 64
XM_005245593.4 XP_005245650.1 7 - 8 770 64
XM_005245594.2 XP_005245651.1 7 - 8 1756 64
XM_006711622.3 XP_006711685.1 7 - 8 2033 64
XM_006711623.3 XP_006711686.1 6 - 7 769 64
XM_006711624.3 XP_006711687.1 6 - 7 796 64
XM_006711625.3 XP_006711688.1 6 - 7 702 64
XM_006711626.2 XP_006711689.1 6 - 7 1688 64
XM_011510136.2 XP_011508438.2 11 - 12 1451 64
XM_011510137.2 XP_011508439.2 11 - 12 1454 64
XM_011510139.2 XP_011508441.1 11 - 12 1454 64
XM_011510141.2 XP_011508443.1 7 - 8 1572 64
XM_011510143.2 XP_011508445.1 3 - 4 318 64
XM_017002794.1 XP_016858283.1 11 - 12 1451 64
XM_017002795.1 XP_016858284.1 7 - 8 786 64
XM_017002796.1 XP_016858285.1 7 - 8 2030 64
XM_017002797.1 XP_016858286.1 7 - 8 770 64
XM_017002798.1 XP_016858287.1 6 - 7 1965 64
XM_017002799.1 XP_016858288.1 6 - 7 766 64
GenBank mRNA AB014551.3 - 11 - 12 1110 64
AF486838.1 - 7 - 8 721 64
AK295924.1 - 7 - 8 1039 64
AK304548.1 - 10 - 11 1179 64
AL512715.1 - 7 - 8 752 64
BC020567.1 - 7 - 8 824 64
BT007407.1 - 6 - 7 643 64

View Full Product Details