Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGEF2
Entrez Gene ID
9181
Gene Name
Rho/Rac guanine nucleotide exchange factor 2
Gene Aliases
GEF, GEF-H1, GEFH1, LFP40, P40
Chromosome Location
Chr.1: 155946839 - 155991263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162383.1 NP_001155855.1 9 - 10 1217 69
NM_001162384.1 NP_001155856.1 9 - 10 1214 69
NM_004723.3 NP_004714.2 9 - 10 1142 69
XM_005245587.2 XP_005245644.1 9 - 10 1270 69
XM_005245588.2 XP_005245645.1 9 - 10 1267 69
XM_005245589.4 XP_005245646.1 9 - 10 1423 69
XM_005245590.3 XP_005245647.1 9 - 10 1145 69
XM_005245591.3 XP_005245648.1 10 - 11 1261 69
XM_005245592.2 XP_005245649.1 9 - 10 1414 69
XM_005245593.4 XP_005245650.1 9 - 10 1145 69
XM_005245594.2 XP_005245651.1 9 - 10 2131 69
XM_006711622.3 XP_006711685.1 9 - 10 2408 69
XM_006711623.3 XP_006711686.1 8 - 9 1144 69
XM_006711624.3 XP_006711687.1 8 - 9 1171 69
XM_006711625.3 XP_006711688.1 8 - 9 1077 69
XM_006711626.2 XP_006711689.1 8 - 9 2063 69
XM_011510136.2 XP_011508438.2 13 - 14 1826 69
XM_011510137.2 XP_011508439.2 13 - 14 1829 69
XM_011510139.2 XP_011508441.1 13 - 14 1829 69
XM_011510141.2 XP_011508443.1 9 - 10 1947 69
XM_011510143.2 XP_011508445.1 5 - 6 693 69
XM_017002794.1 XP_016858283.1 13 - 14 1826 69
XM_017002795.1 XP_016858284.1 9 - 10 1161 69
XM_017002796.1 XP_016858285.1 9 - 10 2405 69
XM_017002797.1 XP_016858286.1 9 - 10 1145 69
XM_017002798.1 XP_016858287.1 8 - 9 2340 69
XM_017002799.1 XP_016858288.1 8 - 9 1141 69
GenBank mRNA AB014551.3 - 13 - 14 1485 69
AF486838.1 - 9 - 10 1096 69
AK295924.1 - 9 - 10 1414 69
AK304548.1 - 12 - 13 1554 69
AL512715.1 - 9 - 10 1127 69
BC020567.1 - 9 - 10 1199 69
BT007407.1 - 8 - 9 1018 69
U72206.1 - 9 - 10 1204 69

View Full Product Details