The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015120.4 | NP_055935.4 | 16 - 17 | 11662 | 103 |
GenBank mRNA | AB002326.3 | - | 9 - 10 | 5351 | 103 |
AJ417593.2 | - | 16 - 17 | 11662 | 103 | |
AX751581.1 | - | 15 - 16 | 11536 | 103 | |
BX647706.1 | - | 9 - 10 | 5575 | 103 |