Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAPTM4B
Entrez Gene ID
55353
Gene Name
lysosomal protein transmembrane 4 beta
Gene Aliases
LAPTM4beta, LC27
Chromosome Location
Chr.8: 97775581 - 97852602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018407.4 NP_060877.3 5 - 6 935 102
GenBank mRNA AF317417.1 - 5 - 6 506 102
AF527412.1 - 5 - 6 864 102
AK075326.1 - 5 - 6 697 102
AL136942.1 - 5 - 6 689 102
AY057051.2 - 5 - 6 935 102
AY261384.1 - 5 - 6 954 102
BC014129.2 - 5 - 6 691 102
BC031021.1 - 5 - 6 716 102
BC038117.1 - 5 - 6 1483 102

View Full Product Details