Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEI1
Entrez Gene ID
150365
Gene Name
meiotic double-stranded break formation protein 1
Gene Aliases
SPATA38
Chromosome Location
Chr.22: 41699503 - 41799456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152513.3 NP_689726.3 27 - 28 3453 98
XM_011529935.1 XP_011528237.1 28 - 29 3471 98
XM_011529936.1 XP_011528238.1 27 - 28 3447 98
XM_011529937.1 XP_011528239.1 26 - 27 3402 98
XM_011529938.1 XP_011528240.1 26 - 27 3390 98
XM_011529939.1 XP_011528241.1 27 - 28 3507 98
XM_011529940.1 XP_011528242.1 26 - 27 3357 98
XM_011529941.1 XP_011528243.1 27 - 28 3462 98
XM_011529942.2 XP_011528244.1 27 - 28 3698 98
XM_011529943.1 XP_011528245.1 26 - 27 3231 98
XM_011529944.1 XP_011528246.1 25 - 26 3204 98
XM_011529945.2 XP_011528247.1 27 - 28 3509 98
XM_011529946.1 XP_011528248.1 24 - 25 3075 98
XM_011529947.1 XP_011528249.1 22 - 23 2985 98
XM_011529948.2 XP_011528250.1 23 - 24 2879 98
XM_011529949.1 XP_011528251.1 22 - 23 2816 98
XM_011529952.1 XP_011528254.1 22 - 23 2953 98
XM_011529954.1 XP_011528256.1 17 - 18 2257 98
XM_017028633.1 XP_016884122.1 27 - 28 3402 98
XM_017028635.1 XP_016884124.1 13 - 14 1837 98
GenBank mRNA AK092934.1 - 13 - 14 1822 98
AK094461.1 - 9 - 10 1239 98
AK128158.1 - 11 - 12 6177 98
AK301435.1 - 12 - 13 1659 98
AY952376.1 - 12 - 13 2165 98
AY952377.1 - 11 - 12 2060 98
BC029582.1 - 7 - 8 1026 98
BC032248.1 - 4 - 5 787 98

View Full Product Details