Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEI1
Entrez Gene ID
150365
Gene Name
meiotic double-stranded break formation protein 1
Gene Aliases
SPATA38
Chromosome Location
Chr.22: 41699503 - 41799456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152513.3 NP_689726.3 28 - 29 3558 89
XM_011529935.1 XP_011528237.1 29 - 30 3576 89
XM_011529936.1 XP_011528238.1 28 - 29 3552 89
XM_011529937.1 XP_011528239.1 27 - 28 3507 89
XM_011529938.1 XP_011528240.1 27 - 28 3495 89
XM_011529940.1 XP_011528242.1 27 - 28 3462 89
XM_011529942.2 XP_011528244.1 28 - 29 3803 89
XM_011529943.1 XP_011528245.1 27 - 28 3336 89
XM_011529944.1 XP_011528246.1 26 - 27 3309 89
XM_011529945.2 XP_011528247.1 28 - 29 3614 89
XM_011529946.1 XP_011528248.1 25 - 26 3180 89
XM_011529947.1 XP_011528249.1 23 - 24 3090 89
XM_011529948.2 XP_011528250.1 24 - 25 2984 89
XM_011529949.1 XP_011528251.1 23 - 24 2921 89
XM_011529952.1 XP_011528254.1 23 - 24 3058 89
XM_011529954.1 XP_011528256.1 18 - 19 2362 89
XM_017028633.1 XP_016884122.1 28 - 29 3507 89
XM_017028635.1 XP_016884124.1 14 - 15 1942 89
GenBank mRNA AK092934.1 - 14 - 15 1927 89
AK093341.1 - 7 - 8 1523 89
AK094461.1 - 10 - 11 1344 89
AK097834.1 - 8 - 9 1186 89
AK128158.1 - 12 - 13 6282 89
AK301435.1 - 13 - 14 1764 89
AY952376.1 - 13 - 14 2270 89
AY952377.1 - 12 - 13 2165 89
BC029582.1 - 8 - 9 1131 89
BC070111.1 - 23 - 24 3668 89

View Full Product Details