The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257189.1 | NP_001244118.1 | 10 - 11 | 1898 | 68 |
NM_019082.3 | NP_061955.1 | 11 - 12 | 2018 | 68 | |
GenBank mRNA | AF247666.1 | - | 11 - 12 | 1408 | 68 |
AJ131712.1 | - | 11 - 12 | 1442 | 68 | |
AK125595.1 | - | 11 - 12 | 2564 | 68 | |
AK222662.1 | - | 11 - 12 | 1407 | 68 | |
AK315363.1 | - | 10 - 11 | 1287 | 68 | |
AL136700.1 | - | 11 - 12 | 1384 | 68 | |
BC001235.1 | - | 11 - 12 | 1405 | 68 | |
CR457213.1 | - | 11 - 12 | 1375 | 68 |