Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THNSL1
Entrez Gene ID
79896
Gene Name
threonine synthase like 1
Gene Aliases
TSH1
Chromosome Location
Chr.10: 24952319 - 25026664 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024838.4 NP_079114.3 2 - 3 275 120
XM_005252597.2 XP_005252654.1 2 - 3 351 120
XM_017016665.1 XP_016872154.1 3 - 4 361 120
GenBank mRNA AK125249.1 - 2 - 3 203 120
BC035132.2 - 2 - 3 196 120
BX647989.1 - 2 - 3 188 120
DA496769.1 - 2 - 3 288 120

View Full Product Details