Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUCKS1
Entrez Gene ID
64710
Gene Name
nuclear casein kinase and cyclin dependent kinase substrate 1
Gene Aliases
JC7, NUCKS
Chromosome Location
Chr.1: 205712819 - 205750244 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022731.4 NP_073568.2 6 - 7 805 63
XM_005245453.1 XP_005245510.1 6 - 7 778 63
GenBank mRNA AB049824.1 - 6 - 7 736 63
AJ012584.1 - 6 - 7 632 63
AK025133.1 - 6 - 7 600 63
AK225228.1 - 6 - 7 600 63
AY823399.1 - 5 - 6 455 63
BC000805.1 - 6 - 7 726 63

View Full Product Details