The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173461.1 | NP_001166932.1 | 3 - 4 | 954 | 72 |
NM_001173462.1 | NP_001166933.1 | 4 - 5 | 1050 | 72 | |
NM_207386.3 | NP_997269.2 | 5 - 6 | 1107 | 72 | |
XM_017024618.1 | XP_016880107.1 | 4 - 5 | 1011 | 72 | |
GenBank mRNA | AK127379.1 | - | 3 - 4 | 648 | 72 |
AK128003.1 | - | 4 - 5 | 450 | 72 | |
DQ070854.1 | - | 3 - 4 | 72 |