The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005270498.4 | XP_005270555.1 | 9 - 10 | 1802 | 126 |
XM_006710379.3 | XP_006710442.1 | 9 - 10 | 943 | 126 | |
XM_011540764.2 | XP_011539066.1 | 10 - 11 | 1743 | 126 | |
XM_017000379.1 | XP_016855868.1 | 7 - 8 | 570 | 126 | |
XM_017000380.1 | XP_016855869.1 | 8 - 9 | 614 | 126 | |
XM_017000381.1 | XP_016855870.1 | 7 - 8 | 523 | 126 | |
XM_017000382.1 | XP_016855871.1 | 6 - 7 | 436 | 126 | |
XM_017000383.1 | XP_016855872.1 | 7 - 8 | 529 | 126 | |
XM_017000384.1 | XP_016855873.1 | 6 - 7 | 427 | 126 | |
XM_017000385.1 | XP_016855874.1 | 5 - 6 | 396 | 126 |