The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000414.3 | NP_000405.1 | 13 - 14 | 1340 | 103 |
NM_001199291.2 | NP_001186220.1 | 14 - 15 | 1539 | 103 | |
NM_001199292.1 | NP_001186221.1 | 12 - 13 | 1286 | 103 | |
NM_001292027.1 | NP_001278956.1 | 14 - 15 | 1405 | 103 | |
NM_001292028.1 | NP_001278957.1 | 13 - 14 | 1465 | 103 | |
GenBank mRNA | AK094049.1 | - | 13 - 14 | 1209 | 103 |
AK293254.1 | - | 13 - 14 | 1212 | 103 | |
AK295440.1 | - | 8 - 9 | 952 | 103 | |
AK298075.1 | - | 14 - 15 | 1467 | 103 | |
AK299685.1 | - | 14 - 15 | 1310 | 103 | |
AK301212.1 | - | 12 - 13 | 1232 | 103 | |
AK308968.1 | - | 9 - 10 | 1054 | 103 | |
AK312449.1 | - | 13 - 14 | 1286 | 103 | |
BC003098.1 | - | 13 - 14 | 1274 | 103 | |
KU178135.1 | - | 13 - 14 | 1207 | 103 | |
X87176.1 | - | 13 - 14 | 1255 | 103 |