Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSD17B4
Entrez Gene ID
3295
Gene Name
hydroxysteroid 17-beta dehydrogenase 4
Gene Aliases
DBP, MFE-2, MPF-2, PRLTS1, SDR8C1
Chromosome Location
Chr.5: 119452443 - 119542335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000414.3 NP_000405.1 16 - 17 1569 103
NM_001199291.2 NP_001186220.1 17 - 18 1768 103
NM_001199292.1 NP_001186221.1 15 - 16 1515 103
NM_001292027.1 NP_001278956.1 17 - 18 1634 103
NM_001292028.1 NP_001278957.1 16 - 17 1694 103
GenBank mRNA AK094049.1 - 16 - 17 1438 103
AK293254.1 - 16 - 17 1441 103
AK295440.1 - 11 - 12 1181 103
AK298075.1 - 17 - 18 1696 103
AK299685.1 - 17 - 18 1539 103
AK301212.1 - 15 - 16 1461 103
AK312449.1 - 16 - 17 1515 103
BC003098.1 - 16 - 17 1503 103
KU178135.1 - 16 - 17 1436 103
X87176.1 - 16 - 17 1484 103

View Full Product Details