Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHODL
Entrez Gene ID
140578
Gene Name
chondrolectin
Gene Aliases
C21orf68, MT75, PRED12
Chromosome Location
Chr.21: 17819329 - 18267371 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204174.1 NP_001191103.1 2 - 3 404 62
NM_001204175.1 NP_001191104.1 2 - 3 370 62
NM_001204176.1 NP_001191105.1 3 - 4 470 62
NM_001204177.1 NP_001191106.1 2 - 3 370 62
NM_001204178.1 NP_001191107.1 3 - 4 470 62
NM_024944.2 NP_079220.2 2 - 3 779 62
XM_011529453.2 XP_011527755.1 2 - 3 1013 62
XM_011529457.2 XP_011527759.1 3 - 4 776 62
XM_017028273.1 XP_016883762.1 2 - 3 399 62
GenBank mRNA AF257472.1 - 2 - 3 779 62
AF523313.1 - 3 - 4 469 62
AF523314.1 - 3 - 4 470 62
AF523315.1 - 2 - 3 371 62
AK022689.1 - 3 - 4 652 62
AK296009.1 - 2 - 3 404 62
AK313725.1 - 2 - 3 427 62
AY358608.1 - 2 - 3 765 62
BC009418.1 - 2 - 3 749 62

View Full Product Details