The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052863.2 | NP_443095.2 | 1 - 2 | 81 | 86 |
GenBank mRNA | AF086152.1 | - | 1 - 2 | 77 | 86 |
AF313458.1 | - | 1 - 2 | 66 | 86 | |
AF436839.1 | - | 1 - 2 | 77 | 86 | |
AY359064.1 | - | 1 - 2 | 134 | 86 | |
BC029176.1 | - | 1 - 2 | 71 | 86 | |
BC072673.1 | - | 1 - 2 | 77 | 86 |