Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICE1
Entrez Gene ID
23379
Gene Name
interactor of little elongation complex ELL subunit 1
Gene Aliases
KIAA0947
Chromosome Location
Chr.5: 5422673 - 5490234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015325.2 NP_056140.1 15 - 16 6466 91
XM_011513999.2 XP_011512301.1 15 - 16 6466 91
XM_017009285.1 XP_016864774.1 16 - 17 6555 91
GenBank mRNA AB023164.3 - 15 - 16 6445 91
AK024400.1 - 3 - 4 3396 91
AK025279.1 - 2 - 3 232 91
AK130025.1 - 4 - 5 671 91
AL137260.1 - 3 - 4 368 91
AL832204.1 - 3 - 4 4937 91
BC004902.2 - 3 - 4 990 91
BX538020.1 - 3 - 4 3955 91
CR749441.1 - 8 - 9 5737 91

View Full Product Details