The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015325.2 | NP_056140.1 | 16 - 17 | 6661 | 147 |
XM_017009285.1 | XP_016864774.1 | 17 - 18 | 6750 | 147 | |
GenBank mRNA | AB023164.3 | - | 16 - 17 | 6640 | 147 |
AK024400.1 | - | 4 - 5 | 3591 | 147 | |
AK025279.1 | - | 3 - 4 | 427 | 147 | |
AK130025.1 | - | 5 - 6 | 866 | 147 | |
AL137260.1 | - | 4 - 5 | 563 | 147 | |
AL832204.1 | - | 4 - 5 | 5132 | 147 | |
BC004902.2 | - | 4 - 5 | 1185 | 147 | |
BX538020.1 | - | 4 - 5 | 4150 | 147 | |
CR749441.1 | - | 9 - 10 | 5932 | 147 |