The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015325.2 | NP_056140.1 | 17 - 18 | 6764 | 146 |
XM_017009285.1 | XP_016864774.1 | 18 - 19 | 6853 | 146 | |
GenBank mRNA | AB023164.3 | - | 17 - 18 | 6743 | 146 |
AK024400.1 | - | 5 - 6 | 3694 | 146 | |
AK025279.1 | - | 4 - 5 | 530 | 146 | |
AK130025.1 | - | 6 - 7 | 969 | 146 | |
AL137260.1 | - | 5 - 6 | 666 | 146 | |
AL832204.1 | - | 5 - 6 | 5235 | 146 | |
BC004902.2 | - | 5 - 6 | 1288 | 146 | |
BX538020.1 | - | 5 - 6 | 4253 | 146 |