The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320821.1 | NP_001307750.1 | 25 - 26 | 3029 | 75 |
NM_001320822.1 | NP_001307751.1 | 23 - 24 | 2551 | 75 | |
NM_007368.3 | NP_031394.2 | 23 - 24 | 2563 | 75 | |
XM_011534841.2 | XP_011533143.1 | 23 - 24 | 2628 | 75 | |
XM_011534842.2 | XP_011533144.1 | 23 - 24 | 2688 | 75 | |
XM_017020438.1 | XP_016875927.1 | 25 - 26 | 2977 | 75 | |
XM_017020439.1 | XP_016875928.1 | 24 - 25 | 2774 | 75 | |
GenBank mRNA | AK128094.1 | - | 25 - 26 | 2998 | 75 |
AK289746.1 | - | 23 - 24 | 2469 | 75 | |
AK294913.1 | - | 23 - 24 | 2536 | 75 | |
AK313795.1 | - | 23 - 24 | 2497 | 75 | |
BC038456.1 | - | 23 - 24 | 2560 | 75 | |
BC047242.1 | - | 23 - 24 | 2551 | 75 | |
X89399.1 | - | 23 - 24 | 2475 | 75 |