The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320822.1 | NP_001307751.1 | 2 - 3 | 298 | 74 |
NM_007368.3 | NP_031394.2 | 2 - 3 | 310 | 74 | |
XM_011534841.2 | XP_011533143.1 | 2 - 3 | 375 | 74 | |
XM_011534842.2 | XP_011533144.1 | 2 - 3 | 435 | 74 | |
GenBank mRNA | AK289746.1 | - | 2 - 3 | 216 | 74 |
AK294913.1 | - | 2 - 3 | 283 | 74 | |
AK295861.1 | - | 2 - 3 | 239 | 74 | |
BC038456.1 | - | 2 - 3 | 307 | 74 | |
BC047242.1 | - | 2 - 3 | 298 | 74 | |
DA328760.1 | - | 2 - 3 | 311 | 74 | |
DC333077.1 | - | 2 - 3 | 298 | 74 | |
X89399.1 | - | 2 - 3 | 222 | 74 |